Di-tert-butyl-3,3'-Iminodipropionate ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3,3'-Iminodipropionic Acid Di-tert-butyl Ester, Di-tert-butyl 3,3'-azanediyldipropanoate

D4110-1G D4110-5G
TCIAD4110-1GEA 815 SEK
TCIAD4110-1G TCIAD4110-5G
Di-tert-butyl-3,3'-Iminodipropionate ≥97.0% (by GC, titration analysis)
Di-tert-butyl-3,3'-Iminodipropionate
Formel: C₁₄H₂₇NO₄
MW: 273,37 g/mol
MDL Number: MFCD07781379
CAS nummer: 128988-04-5

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR