2-(3,4-Dimethoxyphenyl)ethanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol

D4107-25G D4107-5G
TCIAD4107-25GEA 1700 SEK
TCIAD4107-25G TCIAD4107-5G
2-(3,4-Dimethoxyphenyl)ethanol ≥98.0%
2-(3,4-Dimethoxyphenyl)ethanol
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Kokpunkt: 174°C
MDL Number: MFCD00002894
CAS nummer: 7417-21-2
EINECS: 231-032-5

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR