Delapril hydrochloride ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanyl-N-(2,3-dihydroinden-2-yl)glycine Hydrochloride, 2-((S)-N-(2,3-Dihydro-1H-inden-2-yl)-2-(((S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanamido)acetic acid hydrochloride, Delapril HCl

D4082-1G D4082-100MG
TCIAD4082-1GEA 1275 SEK
TCIAD4082-1G TCIAD4082-100MG
Delapril hydrochloride ≥97.0% (by HPLC, titration analysis)
Delapril hydrochloride
Formel: C₂₆H₃₂N₂O₅
MW: 489,01 g/mol
MDL Number: MFCD00884619
CAS nummer: 83435-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145bc3867fb8453450016a66cbac2fe63370035a&vid=145bc3867fb8453450016a66cbac2fe63370035a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 171.0 to 175.0 deg-C
Specific rotation [a]20/D +17.0 to +20.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR