2,5-Dibromoselenophene ≥98.0%

Leverantör: TCI
Fara

D4028-1G D4028-5G
TCIAD4028-1GEA 1300 SEK
TCIAD4028-1G TCIAD4028-5G
2,5-Dibromoselenophene ≥98.0%
Formel: C₄H₂Br₂Se
Kokpunkt: 104°C
Densitet: 2,50
Storage Temperature: Kylskåp
CAS nummer: 1755-36-8
UN: 3440
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR