4-(2,4-Difluorobenzoyl)piperidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

D3979-25G D3979-5G
TCIAD3979-25GEA 2025 SEK
TCIAD3979-25G TCIAD3979-5G
4-(2,4-Difluorobenzoyl)piperidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-(2,4-Difluorobenzoyl)piperidine hydrochloride
Formel: C₁₂H₁₃F₂NO
MDL Number: MFCD01313310
CAS nummer: 106266-04-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR