3,4'-Dihexyl-2,2'-bithiophene ≥96.0%
Leverantör: TCI EUROPE
D3928-1G D3928-5G
TCIAD3928-1GEA 2075 SEK
TCIAD3928-1G TCIAD3928-5G
3,4'-Dihexyl-2,2'-bithiophene ≥96.0%
3,4'-Dihexyl-2,2'-bithiophene
Formel: C₂₀H₃₀S₂
MW: 334,58 g/mol
Kokpunkt: 165°C
Storage Temperature: Kylskåp
MDL Number: MFCD15072161
CAS nummer: 135926-93-1
Order Now
Specifikation testresultat
Appearance White to Amber to Dark green clear liquid
Purity (GC) min. 96.0 %