2,4-Dibromopyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D3862-5G
D3862-25G
TCIAD3862-5GEA
1050
SEK
TCIAD3862-5G
TCIAD3862-25G
2,4-Dibromopyridine ≥98.0%
2,4-Dibromopyridine
Formel:
C₅H₃Br₂N MW: 236,89 g/mol Kokpunkt: 238°C |
MDL Number:
MFCD01859720 CAS nummer: 58530-53-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...