1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide ≥95.0%

Leverantör: TCI EUROPE
Varning

D3846-1G D3846-5G
TCIAD3846-1GEA 1050 SEK
TCIAD3846-1G TCIAD3846-5G
1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide ≥95.0%
1,3-Di-tert-butyl-1,3,2-diazaphospholidine-2-oxide
Formel: C₁₀H₂₃N₂OP
MW: 218,28 g/mol
CAS nummer: 854929-38-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR