2,4-Dimethyloxazole ≥98.0%
Leverantör: TCI EUROPE
Varning
D3766-1G D3766-5G
TCIAD3766-1GEA 1375 SEK
TCIAD3766-1G TCIAD3766-5G
2,4-Dimethyloxazole ≥98.0%
2,4-Dimethyloxazole
Formel: C₅H₇NO
MW: 97,12 g/mol
Kokpunkt: 108°C
MDL Number: MFCD00234124
CAS nummer: 7208-05-1
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=216391fa3808ede531c5a7b934c99b3f406ae1a0&vid=216391fa3808ede531c5a7b934c99b3f406ae1a0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %