2,5-Dimethyl-3-furanthiol ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Mercapto-2,5-dimethylfuran, 2,5-Dimethylfuran-3-thiol

D3764-1G D3764-5G
TCIAD3764-1GEA 830 SEK
TCIAD3764-1G TCIAD3764-5G
2,5-Dimethyl-3-furanthiol ≥97.0%
2,5-Dimethyl-3-furanthiol
Formel: C₆H₈OS
MW: 128,19 g/mol
Kokpunkt: 150°C
MDL Number: MFCD00801026
CAS nummer: 55764-23-3
EINECS: 259-800-5
UN: 3336
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR