2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98.0%

Leverantör: TCI EUROPE

Synonyms: D-Fructopyranose diacetonide, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methanol, Diacetonefructose

D3758-25G D3758-5G
TCIAD3758-25GEA 1675 SEK
TCIAD3758-25G TCIAD3758-5G
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98.0%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
Formel: C₁₂H₂₀O₆
MW: 260,29 g/mol
Storage Temperature: Frys
MDL Number: MFCD00022183
CAS nummer: 20880-92-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 94.0 to 98.0 deg-C
Specific rotation [a]20/D -33.0 to -35.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR