Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Decahydro-2,6-naphthalenedicarboxylic Acid Dimethyl Ester

D3745-25G D3745-5G
TCIAD3745-25GEA 9125 SEK
TCIAD3745-25G TCIAD3745-5G
Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) ≥98.0%
Dimethyl decahydro-2,6-naphthalenedicarboxylate (mixture of isomers)
Formel: C₁₄H₂₂O₄
MW: 254,33 g/mol
Kokpunkt: 140°C
CAS nummer: 3068-02-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR