3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3,6-Diphenylpyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione, Pigment Red 255

D3743-25G D3743-5G
TCIAD3743-25GEA 3650 SEK
TCIAD3743-25G TCIAD3743-5G
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0%
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione
Formel: C₁₈H₁₂N₂O₂
MW: 288,31 g/mol
Smältpunkt: 360°C
MDL Number: MFCD11870113
CAS nummer: 54660-00-3

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR