2,5-Dibromothieno[3,2-b]thiophene ≥98.0%

Leverantör: TCI EUROPE

D3678-1G D3678-200MG D3678-5G
TCIAD3678-1GEA 1025 SEK
TCIAD3678-1G TCIAD3678-200MG TCIAD3678-5G
2,5-Dibromothieno[3,2-b]thiophene ≥98.0%
2,5-Dibromothieno[3,2-b]thiophene
Formel: C₆H₂Br₂S₂
MW: 298,0181 g/mol
Storage Temperature: Frys
MDL Number: MFCD03931293
CAS nummer: 25121-87-3

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR