2,6-Dimethylpyridin-4-ol ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2,6-Dimethyl-4-pyridinol, 2,6-Dimethyl-4-hydroxypyridine
D3638-1G D3638-5G
TCIAD3638-1GEA 670 SEK
TCIAD3638-1G TCIAD3638-5G
2,6-Dimethylpyridin-4-ol ≥98.0%
2,6-Dimethylpyridin-4-ol
Formel: C₇H₉NO
MW: 123,1525 g/mol
Kokpunkt: 351°C
MDL Number: MFCD00956021
CAS nummer: 13603-44-6
EINECS: 237-089-2
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 228.0 to 232.0 deg-C