3,5-Dichloro-2,4-difluoroaniline ≥94.0%
Leverantör: TCI
|
Varning
|
D3637-25G
D3637-5G
TCIAD3637-25GEA
1550
SEK
TCIAD3637-25G
TCIAD3637-5G
3,5-Dichloro-2,4-difluoroaniline ≥94.0%
3,5-Dichloro-2,4-difluoroaniline
Formel:
C₆H₃Cl₂F₂N |
MDL Number:
MFCD00044104 CAS nummer: 83121-15-7 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 94.0 % |
Melting point | 72.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...