Dimethyl-4-hydroxyisophthalate ≥97.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 4-Hydroxyisophthalic Acid Dimethyl Ester, Dimethyl 4-hydroxyisophthalate

D3562-1G D3562-5G
TCIAD3562-1GEA 815 SEK
TCIAD3562-1G TCIAD3562-5G
Dimethyl-4-hydroxyisophthalate ≥97.0% (by GC, titration analysis)
Dimethyl-4-hydroxyisophthalate
Formel: C₁₀H₁₀O₅
MW: 210.19 g/mol
MDL Number: MFCD00152444
CAS nummer: 5985-24-0
EINECS: 227-803-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR