2,5-Dimethyl-4-methoxy-3(2H)-furanone ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
4-Methoxy-2,5-dimethylfuran-3(2H)-one, 4-Methoxy-2,5-dimethyl-3(2H)-furanone
D3499-25G
D3499-5G
TCIAD3499-25GEA
775
SEK
TCIAD3499-25G
TCIAD3499-5G
2,5-Dimethyl-4-methoxy-3(2H)-furanone ≥97.0%
2,5-Dimethyl-4-methoxy-3(2H)-furanone
Formel:
C₇H₁₀O₃ MW: 142,15 g/mol Kokpunkt: 63°C Flampunkt: 75°C |
MDL Number:
MFCD00209504 CAS nummer: 4077-47-8 EINECS: 223-797-9 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...