2,5-Diacetoxyhexane ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2,5-Hexanediol Diacetate
D3496-25G
D3496-5G
TCIAD3496-25GEA
4250
SEK
TCIAD3496-25G
TCIAD3496-5G
2,5-Diacetoxyhexane ≥98.0%
2,5-Diacetoxyhexane
Formel:
C₁₀H₁₈O₄ MW: 202,25 g/mol Kokpunkt: 122°C |
CAS nummer:
10299-35-1 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...