(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

D3444-1G D3444-100MG
TCIAD3444-1GEA 5175 SEK
TCIAD3444-1G TCIAD3444-100MG
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine
Formel: C₁₆H₁₆N₂
MW: 236,32 g/mol
MDL Number: MFCD09038520
CAS nummer: 181139-49-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C
Specific rotation [a]20/D -16.0 to -20.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR