9,9-Dimethylfluorene ≥98.0%
Leverantör: TCI
Synonyms: 9,9-Dimethyl-9H-fluorene
D3235-5G D3235-1G
TCIAD3235-5GEA 1150 SEK
TCIAD3235-5G TCIAD3235-1G
9,9-Dimethylfluorene ≥98.0%
9,9-Dimethylfluorene
Formel: C₁₅H₁₄
Kokpunkt: 287°C
MDL Number: MFCD00114670
CAS nummer: 4569-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfc75e928257bcf10478481dc4272d59cd265868&vid=bfc75e928257bcf10478481dc4272d59cd265868&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C