Dihydrojasmonic acid ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-(Carboxymethyl)-2-pentylcyclopentanone, 2-Amyl-3-(carboxymethyl)cyclopentanone, 3-Oxo-2-pentylcyclopentaneacetic Acid, 2-Amyl-3-oxocyclopentaneacetic Acid

D3225-1G D3225-5G
TCIAD3225-1GEA 1925 SEK
TCIAD3225-1G TCIAD3225-5G
Dihydrojasmonic acid ≥98.0%
Dihydrojasmonic acid
Formel: C₁₂H₂₀O₃
MW: 212,29 g/mol
Storage Temperature: Frys
CAS nummer: 3572-64-3
EINECS: 222-687-8

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR