Diethyl-3-hydroxyglutarate 95,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: 3-Hydroxyglutaric Acid Diethyl Ester, Diethyl 3-hydroxyglutarate

D3196-25G D3196-5G
TCIAD3196-25GEA 845 SEK
TCIAD3196-25G TCIAD3196-5G
Diethyl-3-hydroxyglutarate 95,0 GC_ANALYSMETOD
Diethyl-3-hydroxyglutarate
Formel: C₉H₁₆O₅
MW: 204.22 g/mol
Kokpunkt: 107°C
MDL Number: MFCD00009202
CAS nummer: 32328-03-3
EINECS: 250-992-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR