4,4'-Dimethoxy-trans-stilbene ≥98.0%

Leverantör: TCI EUROPE

D3104-500MG D3104-1G
TCIAD3104-500MGEA 1300 SEK
TCIAD3104-500MG TCIAD3104-1G
4,4'-Dimethoxy-trans-stilbene ≥98.0%
4,4'-Dimethoxy-trans-stilbene
Formel: C₁₆H₁₆O₂
Storage Temperature: Kylskåp
CAS nummer: 15638-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=999195d4d1f2a89b66020e1d972b2caa81363cd6&vid=999195d4d1f2a89b66020e1d972b2caa81363cd6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 213.0 to 217.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR