3,6-Dibromo-9-ethylcarbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,6-Dibromo-9-ethyl-9H-carbazole
D2982-1G
D2982-5G
TCIAD2982-1GEA
805
SEK
TCIAD2982-1G
TCIAD2982-5G
3,6-Dibromo-9-ethylcarbazole ≥98.0%
3,6-Dibromo-9-ethylcarbazole
Formel:
C₁₄H₁₁Br₂N MW: 353,06 g/mol |
MDL Number:
MFCD00138134 CAS nummer: 33255-13-9 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...