3,5-Diacetoxybenzoic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D2844-1G
D2844-5G
TCIAD2844-1GEA
1325
SEK
TCIAD2844-1G
TCIAD2844-5G
3,5-Diacetoxybenzoic acid ≥98.0%
3,5-Diacetoxybenzoic acid
Formel:
C₁₁H₁₀O₆ MW: 238,20 g/mol |
MDL Number:
MFCD00017591 CAS nummer: 35354-29-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...