(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene

D2801-1G D2801-200MG
TCIAD2801-1GEA 3600 SEK
TCIAD2801-1G TCIAD2801-200MG
(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%
(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl
Formel: C₂₂H₁₆Br₂O₂
MW: 472,18 g/mol
MDL Number: MFCD04038415
CAS nummer: 75714-59-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 180.0 to 185.0 deg-C
Specific rotation [a]20/D +10.0 to +14.0 deg (C=1, CHCl3)
Solubility in Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR