1-Dimethylamino-2-propyne ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: N,N-Dimethylpropargylamine, N,N-Dimethyl-2-propyn-1-amine

D2794-25ML D2794-5ML
TCIAD2794-25MLEA 3050 SEK
TCIAD2794-25ML TCIAD2794-5ML
1-Dimethylamino-2-propyne ≥98.0% (by GC, titration analysis)
1-Dimethylamino-2-propyne
Formel: C₅H₉N
MW: 83,13 g/mol
Flampunkt: –7°C
MDL Number: MFCD00008575
CAS nummer: 7223-38-3
EINECS: 230-620-9
UN: 2733
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 4.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR