6-(Dimethylamino)fulvene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-(Dimethylaminomethylene)-1,3-cyclopentadiene
D2785-1G
D2785-5G
TCIAD2785-1GEA
1300
SEK
TCIAD2785-1G
TCIAD2785-5G
6-(Dimethylamino)fulvene ≥98.0%
6-(Dimethylamino)fulvene
Formel:
C₈H₁₁N MW: 121,18 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00013275 CAS nummer: 696-68-4 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...