ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97.0%
Leverantör: TCI
Varning
Synonyms: Dihydro-3,3-dimethyl-2(3H)-furanone, α,α-Dimethyl-γ-butyrolactone
D2733-25G D2733-5G
TCIAD2733-25GEA 10150 SEK
TCIAD2733-25G TCIAD2733-5G
ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97.0%
alpha,alpha-Dimethyl-gamma-butyrolactone
Formel: C₆H₁₀O₂
MW: 114.14 g/mol
Kokpunkt: 194°C
Smältpunkt: 6°C
MDL Number: MFCD03844788
CAS nummer: 3709-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ea32bdada66f6688bce80228711c0413709273e&vid=7ea32bdada66f6688bce80228711c0413709273e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %