(+)-3,9-Dibromocamphor ≥97.0%

Leverantör: TCI EUROPE
Varning

D2715-25G D2715-5G
TCIAD2715-25GEA 7725 SEK
TCIAD2715-25G TCIAD2715-5G
(+)-3,9-Dibromocamphor ≥97.0%
(+)-3,9-Dibromocamphor
Formel: C₁₀H₁₄Br₂O
MW: 310,03 g/mol
MDL Number: MFCD00167983
CAS nummer: 10293-10-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Specific rotation [a]20/D +95.0 to +105.0 deg (C=1,chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR