(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
(2R,3R)-2,3-Bis(pivaloyloxy)succinic acid, (-)-O,O'-Dipivaloyl-L-tartaric acid, (-)-Bis(2,2-dimethylpropionyl)-L-tartaric Acid
D2686-25G
D2686-5G
TCIAD2686-25GEA
2675
SEK
TCIAD2686-25G
TCIAD2686-5G
(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
(-)-Dipivaloyl-L-tartaric acid
Formel:
C₁₄H₂₂O₈ MW: 318,32 g/mol |
MDL Number:
MFCD00066535 CAS nummer: 65259-81-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...