2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 2,5-Diaminopyrimidine-4,6-diol Hydrochloride, 2,5-Diamino-6-hydroxy-4(1H)-pyrimidinone Hydrochloride

D2634-25G D2634-5G
TCIAD2634-25GEA 3250 SEK
TCIAD2634-25G TCIAD2634-5G
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride
Formel: C₄H₆N₄O₂
MW: 178,58 g/mol
MDL Number: MFCD00082893
CAS nummer: 56830-58-1

Order Now

Specifikation testresultat

Appearance White to Brown to Dark purple powder to crystal
Purity (Argentmetric Titration) min. 98.0 % (calcd.on dried substance)
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR