2,4'-Dihydroxydiphenyl sulphone ≥98.0%

Leverantör: TCI
Varning

D2567-25G D2567-500G
TCIAD2567-25GEA 270 SEK
TCIAD2567-25G TCIAD2567-500G
2,4'-Dihydroxydiphenyl sulphone ≥98.0%
2,4'-Dihydroxydiphenyl sulphone
Formel: C₁₂H₁₀O₄S
MDL Number: MFCD01631305
CAS nummer: 5397-34-2
EINECS: 226-421-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR