6,6'-Dithiodinicotinic acid 98,0 HPLC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 6,6'-Dithiobis(3-pyridinecarboxylic Acid)

D2564-25G D2564-5G
TCIAD2564-25GEA 5250 SEK
TCIAD2564-25G TCIAD2564-5G
6,6'-Dithiodinicotinic acid 98,0 HPLC_ANALYSMETOD
6,6'-Dithiodinicotinic acid
Formel: C₁₂H₈N₂O₄S₂
MW: 308.33 g/mol
MDL Number: MFCD00006454
CAS nummer: 15658-35-2
EINECS: 239-729-6

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR