(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(S)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulphonate, L-(+)-1,2-Isopropylideneglycerol 3-(p-Toluenesulphonate)
D2550-1G
D2550-5G
TCIAD2550-1GEA
1525
SEK
TCIAD2550-1G
TCIAD2550-5G
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate ≥98.0%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate
Formel:
C₁₃H₁₈O₅S MW: 286,34 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00063234 CAS nummer: 23735-43-5 |
Specifikation testresultat
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +7.0 to +8.0 deg (C=2, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...