3,4-Diaminothiophene dihydrochloride ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Thiophene-3,4-diamine dihydrochloride
D2378-1G
D2378-100MG
TCIAD2378-1GEA
1700
SEK
TCIAD2378-1G
TCIAD2378-100MG
3,4-Diaminothiophene dihydrochloride ≥98.0%
3,4-Diaminothiophene dihydrochloride
Formel:
C₄H₆N₂S MW: 187,08 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00671511 CAS nummer: 90069-81-1 |
Specifikation testresultat
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...