Dodecanedioic dihydrazide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Dodecanedioic Dihydrazide, Dodecanediohydrazide

D2342-25G D2342-500G
TCIAD2342-25GEA 445 SEK
TCIAD2342-25G TCIAD2342-500G
Dodecanedioic dihydrazide ≥98.0%
Dodecanedioic dihydrazide
Formel: C₁₂H₂₆N₄O₂
MW: 258,37 g/mol
MDL Number: MFCD00043619
CAS nummer: 4080-98-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 98.0 %
Melting point 186.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR