4,4'-Dimethoxyoctafluorobiphenyl ≥98.0%

Leverantör: TCI
Varning

D2330-1G D2330-5G
TCIAD2330-1GEA 525 SEK
TCIAD2330-1G TCIAD2330-5G
4,4'-Dimethoxyoctafluorobiphenyl ≥98.0%
4,4'-Dimethoxyoctafluorobiphenyl
Formel: C₁₄H₆F₈O₂
MDL Number: MFCD00039802
CAS nummer: 2200-71-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR