Dimethoxymethylphenylsilane ≥98.0%

Leverantör: TCI
Varning

Synonyms: Methylphenyldimethoxysilane, Dimethoxy(methyl)(phenyl)silane

D2319-100ML D2319-25ML
TCIAD2319-100MLEA 885 SEK
TCIAD2319-100ML TCIAD2319-25ML
Dimethoxymethylphenylsilane ≥98.0%
Dimethoxymethylphenylsilane
Formel: C₉H₁₄O₂Si
Kokpunkt: 199°C
Flampunkt: 82°C
MDL Number: MFCD00008343
CAS nummer: 3027-21-2
EINECS: 221-192-4

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR