2,5-Dimethoxy-1,4-benzoquinone ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2,5-Dimethoxy-p-quinone
D2250-25G
D2250-5G
TCIAD2250-25GEA
4575
SEK
TCIAD2250-25G
TCIAD2250-5G
2,5-Dimethoxy-1,4-benzoquinone ≥98.0%
2,5-Dimethoxy-1,4-benzoquinone
Formel:
C₈H₈O₄ MW: 168,15 g/mol |
MDL Number:
MFCD00189386 CAS nummer: 3117-03-1 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...