2,5-Dibromo-1,4-benzoquinone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,5-Dibromocyclohexa-2,5-diene-1,4-dione, 2,5-Dibromo-p-quinone
D2249-1G
D2249-5G
TCIAD2249-1GEA
1925
SEK
TCIAD2249-1G
TCIAD2249-5G
2,5-Dibromo-1,4-benzoquinone ≥98.0%
2,5-Dibromo-1,4-benzoquinone
Formel:
C₆H₂Br₂O₂ MW: 265,89 g/mol |
MDL Number:
MFCD00156859 CAS nummer: 1633-14-3 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 192.0 to 195.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...