4-Dodecyloxyphthalonitrile ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2-Dicyano-4-dodecyloxybenzene
D2237-1G
D2237-5G
TCIAD2237-1GEA
885
SEK
TCIAD2237-1G
TCIAD2237-5G
4-Dodecyloxyphthalonitrile ≥99.0%
4-Dodecyloxyphthalonitrile
Formel:
C₂₀H₂₈N₂O MW: 312,46 g/mol |
MDL Number:
MFCD00191413 CAS nummer: 161082-75-3 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 61.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...