2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,2-Dimethyl-7-hydroxycoumaran, 2,2-Dimethyl-2,3-dihydrobenzofuran-7-ol
D2154-25G
D2154-100G
TCIAD2154-25GEA
430
SEK
TCIAD2154-25G
TCIAD2154-100G
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran ≥98.0%
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran
Formel:
C₁₀H₁₂O₂ MW: 164,20 g/mol Kokpunkt: 95°C |
MDL Number:
MFCD00075382 CAS nummer: 1563-38-8 EINECS: 216-350-4 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...