Glycerol-ɑ,ɑ'-diallyl ether ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: GDAE, 1,3-Diallyloxy-2-propanol

D2146-25ML D2146-500ML
TCIAD2146-25MLEA 870 SEK
TCIAD2146-25ML TCIAD2146-500ML
Glycerol-ɑ,ɑ'-diallyl ether ≥98.0%
Glycerol-alpha,alpha'-diallyl ether
Formel: C₉H₁₆O₃
MW: 172,22 g/mol
Kokpunkt: 106°C
MDL Number: MFCD00085245
CAS nummer: 17018-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c490310a3c6e25277962316e404cbcfdb0d5f28&vid=7c490310a3c6e25277962316e404cbcfdb0d5f28&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR