Benzyl viologen dichloride ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1,1'-Dibenzyl-[4,4'-bipyridine]-1,1'-diium chloride, 1,1'-Dibenzyl-4,4'-bipyridinium dichloride
D2137-1G
D2137-25G
D2137-5G
TCIAD2137-1GEA
630
SEK
TCIAD2137-1G
TCIAD2137-25G
TCIAD2137-5G
Benzyl viologen dichloride ≥98.0%
Benzyl viologen dichloride
Formel:
C₂₄H₂₂Cl₂N₂ MW: 409.35 g/mol |
MDL Number:
MFCD00011991 CAS nummer: 1102-19-8 EINECS: 214-158-5 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 90.0 % |
Water | max. 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...