3,6-Diaminocarbazole ≥98.0%

Leverantör: TCI
Fara

D2116-1G D2116-5G
TCIAD2116-1GEA 2725 SEK
TCIAD2116-1G TCIAD2116-5G
3,6-Diaminocarbazole ≥98.0%
3,6-Diaminocarbazole
Formel: C₁₂H₁₁N₃
MDL Number: MFCD00143081
CAS nummer: 86-71-5
EINECS: 201-692-9
UN: 3259
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR