1,2-Diamino-2-methylpropane ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Methyl-1,2-propanediamine, 2-Methylpropane-1,2-diamine
D2102-25ML D2102-5ML
TCIAD2102-25MLEA 5600 SEK
TCIAD2102-25ML TCIAD2102-5ML
1,2-Diamino-2-methylpropane ≥97.0% (by GC, titration analysis)
1,2-Diamino-2-methylpropane
Formel: C₄H₁₂N₂
MW: 88,15 g/mol
Kokpunkt: 52°C
Flampunkt: 23°C
MDL Number: MFCD00008054
CAS nummer: 811-93-8
EINECS: 212-374-4
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28506c2797a65eddbe7e1860ff584807ef3152cc&vid=28506c2797a65eddbe7e1860ff584807ef3152cc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %