1,11-Diaminoundecane 98,0 GC, T
Leverantör: TCI
|
Varning
|
Synonyms:
1,11-Undecanediamine, Undecamethylenediamine
D2075-1G
D2075-5G
TCIAD2075-1GEA
670
SEK
TCIAD2075-1G
TCIAD2075-5G
1,11-Diaminoundecane 98,0 GC, T
1,11-Diaminoundecane
Formel:
C₁₁H₂₆N₂ MW: 186.34 g/mol Kokpunkt: 152°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00142358 CAS nummer: 822-08-2 EINECS: 212-486-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 43.0 to 46.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...