trans-1,2-Di(2-thienyl)ethylene ≥98.0%
Leverantör: TCI EUROPE
D2074-1G
D2074-5G
TCIAD2074-1GEA
1300
SEK
TCIAD2074-1G
TCIAD2074-5G
trans-1,2-Di(2-thienyl)ethylene ≥98.0%
trans-1,2-Di(2-thienyl)ethylene
Formel:
C₁₀H₈S₂ |
MDL Number:
MFCD00145416 CAS nummer: 13640-78-3 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 133.0 to 136.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...